missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloroisonicotinic acid, 98%, ACROS Organics™

Brand:  Acros Organics 373120010

Code : 29

Additional Details : CAS Number : 5398-44-7 Weight : 0.05100kg


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10515663

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2, 6-Dichloroisonicotinic acid
97.5
≥97.5%
C6H3Cl2NO2
MFCD00234147
5398-44-7
100.0
98%
192

Specifications

2, 6-Dichloroisonicotinic acid
206° to 210°C
5398-44-7
100.0
Powder and/or Chunks
98%
192
Glass bottle
Off-white to beige or light brownish
1g
97.5
Authentic
≥97.5%
C6H3Cl2NO2
MFCD00234147
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2,6-Dichloroisonicotinic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS