missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloroindophenol, sodium salt hydrate, 95%, Thermo Scientific Chemicals

Catalog Number 10746161
Click to view available options
Quantity:
1 g
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1266615-56-8
C12H6Cl2NNaO2
290.07
MFCD00150014
CVSUAFOWIXUYQA-UHFFFAOYSA-M
Tillman's reagent hydrate
23696612
[Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1
This item is not returnable. View return policy
1266615-56-8
C12H6Cl2NNaO2
290.07
MFCD00150014
CVSUAFOWIXUYQA-UHFFFAOYSA-M
Tillman's reagent hydrate
23696612
[Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Assay Percent Range 98+%
Solubility Information Solubility in water: soluble. Other solubilities: freely soluble in alcohol
Infrared Spectrum Authentic
IUPAC Name sodium 4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate
Formula Weight 290.07
Loss on Drying 14% max. (1 g, 120°C)
Percent Purity ≥98%
Packaging Glass bottle
Suitability For determination of ascorbic acid: Passes Test
Color Dark Green
Quantity 25 g
Physical Form Powder
Chemical Name or Material 2, 6-Dichloroindophenol, sodium salt hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'