missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzyl chloride, 97%, Thermo Scientific™

Catalog Number 11454060
Click to view available options
Quantity:
25 g
100 g
250 g
This item is not returnable. View return policy
2014-83-7
C7H5Cl3
195.467
MFCD00000897
LBOBESSDSGODDD-UHFFFAOYSA-N
2,6-dichlorobenzyl chloride, 1,3-dichloro-2-chloromethyl benzene, alpha,2,6-trichlorotoluene, 2,6-dichlorobenzylchloride, 1,3-dichloro-2-chloromethyl-benzene, benzene, 1,3-dichloro-2-chloromethyl, toluene, .alpha.,2,6-trichloro, a,2,6-trichlorotoluene, dichlorobenzyl 2,6-chloride, 1,3-ddichloro-2-chloromethyl benzene
74832
1,3-dichloro-2-(chloromethyl)benzene
C1=CC(=C(C(=C1)Cl)CCl)Cl
This item is not returnable. View return policy
2014-83-7
C7H5Cl3
195.467
MFCD00000897
LBOBESSDSGODDD-UHFFFAOYSA-N
2,6-dichlorobenzyl chloride, 1,3-dichloro-2-chloromethyl benzene, alpha,2,6-trichlorotoluene, 2,6-dichlorobenzylchloride, 1,3-dichloro-2-chloromethyl-benzene, benzene, 1,3-dichloro-2-chloromethyl, toluene, .alpha.,2,6-trichloro, a,2,6-trichlorotoluene, dichlorobenzyl 2,6-chloride, 1,3-ddichloro-2-chloromethyl benzene
74832
1,3-dichloro-2-(chloromethyl)benzene
C1=CC(=C(C(=C1)Cl)CCl)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 36°C to 39°C
Boiling Point 117°C to 119°C (14 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Quantity 100 g
UN Number UN3261
Beilstein 472282
Formula Weight 195.48
Percent Purity 97%
Chemical Name or Material 2,6-Dichlorobenzyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'