missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,6-Dichlorobenzyl alcohol, 99%

Brand:  Alfa Aesar™ A12196.09

Code : NEW

Additional Details : CAS Number : 15258-73-8 Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 11454853

  • £14.42 / 10g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2,6-Dichlorobenzyl alcohol
C7H6Cl2O
MFCD00004609
15258-73-8
177.03
1938356

Specifications

2,6-Dichlorobenzyl alcohol
15258-73-8
C7H6Cl2O
MFCD00004609
1938356
10g
99%
177.03
150°C (25mmHg)
95° to 99°C
Videos
SDS