missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-beta-nitrostyrene, 97%, Thermo Scientific™

Catalog Number p-7080544
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
22482-43-5
C8H5Cl2NO2
218.033
MFCD00024821
VXNHQIKJDIOBEC-SNAWJCMRSA-N
1,3-dichloro-2-2-nitrovinyl benzene, 2,6-dichloro-beta-nitrostyrene, 1,3-dichloro-2-2-nitroethenyl benzene, 1-2,6-dichlorophenyl-2-nitroethylene, 1,3-dichloro-2-e-2-nitroethenyl benzene, styrene, 2,6-dichloro-beta-nitro, 1-3,4-methylenedioxy phenyl-2-nitroethene, benzene, 1,3-dichloro-2-2-nitroethenyl, benzene,1,3-dichloro-2-2-nitroethenyl, 2-1e-2-nitrovinyl-1,3-dichlorobenzene
5369223
1,3-dichloro-2-[(E)-2-nitroethenyl]benzene
C1=CC(=C(C(=C1)Cl)C=C[N+](=O)[O-])Cl
This item is not returnable. View return policy
22482-43-5
C8H5Cl2NO2
218.033
MFCD00024821
VXNHQIKJDIOBEC-SNAWJCMRSA-N
1,3-dichloro-2-2-nitrovinyl benzene, 2,6-dichloro-beta-nitrostyrene, 1,3-dichloro-2-2-nitroethenyl benzene, 1-2,6-dichlorophenyl-2-nitroethylene, 1,3-dichloro-2-e-2-nitroethenyl benzene, styrene, 2,6-dichloro-beta-nitro, 1-3,4-methylenedioxy phenyl-2-nitroethene, benzene, 1,3-dichloro-2-2-nitroethenyl, benzene,1,3-dichloro-2-2-nitroethenyl, 2-1e-2-nitrovinyl-1,3-dichlorobenzene
5369223
1,3-dichloro-2-[(E)-2-nitroethenyl]benzene
C1=CC(=C(C(=C1)Cl)C=C[N+](=O)[O-])Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 64°C to 65°C
Quantity 1 g
Beilstein 1960934
Formula Weight 218.04
Percent Purity 97%
Chemical Name or Material 2,6-Dichloro-beta-nitrostyrene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'