missing translation for 'onlineSavingsMsg'
Learn More

(2,6-Dichloro-4-pyridyl)methanol, 95%, Maybridge

Brand:  Maybridge RF01034EA

Code : 31

Additional Details : CAS Number : 101990-69-6 Weight : 0.06000kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 10614205

  • £340.00 / 10g


Estimated Shipment date
from Supplier 23-07-2018
Add to basket

Specifications

Chemical Identifiers

(2, 6-Dichloro-4-pyridyl)methanol
95.0
95%
178.02
101990-69-6
95%
C6H5Cl2NO
MFCD00052638

Specifications

(2, 6-Dichloro-4-pyridyl)methanol
101990-69-6
Amber glass bottle
95%
178.02
10g
95.0
95%
C6H5Cl2NO
MFCD00052638
Videos
SDS