missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloro-4-pyridyl isocyanate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 2,6-dichloro-4-pyridyl isocyanate, 2,6-dichloropyridin-4-isocyanate, 2,6-dichloropyridin-4-yl isocyanate, pyridine,2,6-dichloro-4-isocyanato, pyridine, 2,6-dichloro-4-isocyanato, 2,6-dichloropyridin-4-ylisocyanate, pyridine, 2,6-dichloro-4-isocyanato-9ci |
Formula Weight | 189 |
Percent Purity | 90% |
Chemical Name or Material | 2, 6-Dichloro-4-isocyanatopyridine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'