missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,6-Dichloro-4-nitrophenol, 98%

Brand:  Alfa Aesar™ A10988.36

Code : NEW

Additional Details : CAS Number : 618-80-4 Weight : 0.50000kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 500g

 View more versions of this product

Product Code. 11474433

  • £365.00 / 500g


Estimated Shipment date
from Supplier 29-05-2018
Add to basket

Specifications

Chemical Identifiers

2,6-Dichloro-4-nitrophenol
C6H3Cl2NO3
MFCD00014715
1245045
618-80-4
208
UN2811

Specifications

2,6-Dichloro-4-nitrophenol
618-80-4
C6H3Cl2NO3
MFCD00014715
1245045
500g
98%
208
UN2811
≈124°C (dec.)
Videos
SDS