Learn More
Alfa Aesar™ 2,6-Dichloro-4-nitrophenol, 98%
Brand: Alfa Aesar™ A10988.22
Code : NEW
Additional Details : CAS Number : 618-80-4 Weight : 0.10000kg Transport : UN number : 2811 Chem class : 1 Pack group : III
Quantity | 100g |
---|
Chemical Identifiers
2,6-Dichloro-4-nitrophenol | |
C6H3Cl2NO3 | |
MFCD00014715 | |
1245045 |
618-80-4 | |
208 | |
UN2811 |
Specifications
2,6-Dichloro-4-nitrophenol | |
618-80-4 | |
C6H3Cl2NO3 | |
MFCD00014715 | |
1245045 |
100g | |
98% | |
208 | |
UN2811 | |
≈124°C (dec.) |