missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,6-Dichloro-3-iodopyridine, 98%

Brand:  Alfa Aesar™ H64366.03

Code : NEW

Additional Details : CAS Number : 148493-37-2 Weight : 0.00100kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15451529

  • £11.60 / 1g


Estimated Shipment date
from Supplier 16-02-2018
Add to basket

Description

Description

Applications
It is used as pharmaceutical raw material and intermediate.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
Specifications

Chemical Identifiers

2,6-Dichloro-3-iodopyridine
273.88
UN2811
148493-37-2
MFCD03094686
6135696

Specifications

2,6-Dichloro-3-iodopyridine
White to brown
1g
Powder
273.88
UN2811
Light Sensitive
74° to 79°C
148493-37-2
98%
MFCD03094686
6135696
Videos
SDS