missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-3-fluoroacetophenone, 97%, Thermo Scientific™

Catalog Number p-105137
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
290835-85-7
C8H5Cl2FO
207.03
MFCD02093760
VJBFZHHRVCPAPZ-UHFFFAOYSA-N
2',6'-dichloro-3'-fluoroacetophenone, 1-2,6-dichloro-3-fluorophenyl ethanone, 2,6-dichloro-3-fluoroacetophenone, 1-2,6-dichloro-3-fluorophenyl ethan-1-one, 2,6-dichloro-3-fluro acetophenone, 2,6-dichloro-5-fluoro acetophenone, 2',6'-dichloro-3'-fluroacetophenone, ethanone, 1-2,6-dichloro-3-fluorophenyl, 1-acetyl-2,6-dichloro-3-fluorobenzene, pubchem8532
2733982
1-(2,6-dichloro-3-fluorophenyl)ethanone
CC(=O)C1=C(Cl)C=CC(F)=C1Cl
This item is not returnable. View return policy
290835-85-7
C8H5Cl2FO
207.03
MFCD02093760
VJBFZHHRVCPAPZ-UHFFFAOYSA-N
2',6'-dichloro-3'-fluoroacetophenone, 1-2,6-dichloro-3-fluorophenyl ethanone, 2,6-dichloro-3-fluoroacetophenone, 1-2,6-dichloro-3-fluorophenyl ethan-1-one, 2,6-dichloro-3-fluro acetophenone, 2,6-dichloro-5-fluoro acetophenone, 2',6'-dichloro-3'-fluroacetophenone, ethanone, 1-2,6-dichloro-3-fluorophenyl, 1-acetyl-2,6-dichloro-3-fluorobenzene, pubchem8532
2733982
1-(2,6-dichloro-3-fluorophenyl)ethanone
CC(=O)C1=C(Cl)C=CC(F)=C1Cl
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 250.0°C to 255.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5275 to 1.5295
Quantity 25 g
Formula Weight 207.03
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 6-Dichloro-3-fluoroacetophenone, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dichloro-3-fluoroacetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'