missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-3-fluoroacetophenone, 97%, ACROS Organics™

Brand:  Acros Organics 345511000

Code : 29

Additional Details : CAS Number : 290835-85-7 Weight : 0.15000kg


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10689843

  • £279.80 / 100g


Estimated Shipment
22-06-2018
Add to basket


Specifications

Chemical Identifiers

2, 6-Dichloro-3-fluoroacetophenone
290835-85-7
100.0
97%
207.03
97%
96.0
97%
C8H5Cl2FO
MFCD02093760

Specifications

2, 6-Dichloro-3-fluoroacetophenone
250.0° to 255.0°C
100g
290835-85-7
100.0
Glass bottle
Liquid
97%
207.03
110°C
Clear colorless to light yellow
97%
96.0
Authentic
1.5275 to 1.5295
97%
C8H5Cl2FO
MFCD02093760
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2,6-Dichloro-3-fluoroacetophenone

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS