missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromotoluene, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081736
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
69321-60-4
C7H6Br2
249.93
MFCD00013524
OCSKCBIGEMSDIS-UHFFFAOYSA-N
2,6-dibromotoluene, benzene, 1,3-dibromo-2-methyl, 1,3-dibromo-2-methyl-benzene, pubchem3926, acmc-1bagq, ksc493g1t, #, 2,6-dibromotoluene 1g
34681
1,3-dibromo-2-methylbenzene
CC1=C(Br)C=CC=C1Br
This item is not returnable. View return policy
69321-60-4
C7H6Br2
249.93
MFCD00013524
OCSKCBIGEMSDIS-UHFFFAOYSA-N
2,6-dibromotoluene, benzene, 1,3-dibromo-2-methyl, 1,3-dibromo-2-methyl-benzene, pubchem3926, acmc-1bagq, ksc493g1t, #, 2,6-dibromotoluene 1g
34681
1,3-dibromo-2-methylbenzene
CC1=C(Br)C=CC=C1Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 2°C to 6°C
Density 1.812
Boiling Point 112°C to 113°C (7 mmHg)
Refractive Index 1.606
Quantity 1 g
Beilstein 3235502
Formula Weight 249.94
Percent Purity ≥98%
Chemical Name or Material 2,6-Dibromotoluene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'