missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromopyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-4258931
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
626-05-1
C5H3Br2N
236.89
FEYDZHNIIMENOB-UHFFFAOYSA-N
12274
2,6-dibromopyridine
C1=CC(=NC(=C1)Br)Br
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
626-05-1
C5H3Br2N
236.89
FEYDZHNIIMENOB-UHFFFAOYSA-N
12274
2,6-dibromopyridine
C1=CC(=NC(=C1)Br)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 118.0°C to 119.0°C
Color Gray to White
Flash Point 213°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H3N)Br2
Quantity 25 g
Beilstein 20, V,5, 435
Synonym pyridine, 2,6-dibromo, 2,6-dibromo-pyridine, 2,6-dibromo pyridine, 2,6dibromopyridine, 2,6-dibrompyridine, 2,6 dibromopyridine, 2.6-dibromopyridine, pubchem3204, 2, 6-dibromopyridine, acmc-209n5v
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,ethanol,dioxane,chloroform,,carbon tetrachloride,heptane,aq. naoh,aq. h2so4
Formula Weight 236.89
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 6-Dibromopyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dibromopyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'