missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dibromo-4-(trifluoromethyl)aniline, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 2,6-dibromo-4-trifluoromethyl aniline, 4-amino-3,5-dibromobenzotrifluoride, 2,6-dibromo-4-trifluoromethyl phenylamine, benzenamine,2,6-dibromo-4-trifluoromethyl, benzenamine, 2,6-dibromo-4-trifluoromethyl, pubchem2805, acmc-209ony, buttpark 29\06-99, 2,6-dibromo-4-trifluoromethyl-phenylamine |
Formula Weight | 318.92 |
Percent Purity | 97% |
Chemical Name or Material | 2, 6-Dibromo-4-(trifluoromethyl)aniline |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'