missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-4-methylphenol, 99%, Thermo Scientific Chemicals

Catalog Number 10753904
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
128-37-0
C15H24O
220.35
MFCD00011644
NLZUEZXRPGMBCV-UHFFFAOYSA-N
2,6-di-tert-butyl-4-methylphenol, butylated hydroxytoluene, butylhydroxytoluene, 2,6-di-tert-butyl-p-cresol, 2,6-di-t-butyl-4-methylphenol, ionol, dbpc, dibunol, stavox, bht
31404
CHEBI:34247
2,6-ditert-butyl-4-methylphenol
CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C
128-37-0
C15H24O
220.35
MFCD00011644
NLZUEZXRPGMBCV-UHFFFAOYSA-N
2,6-di-tert-butyl-4-methylphenol, butylated hydroxytoluene, butylhydroxytoluene, 2,6-di-tert-butyl-p-cresol, 2,6-di-t-butyl-4-methylphenol, ionol, dbpc, dibunol, stavox, bht
31404
CHEBI:34247
2,6-ditert-butyl-4-methylphenol
CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 69.0°C to 71.0°C
Color White
Boiling Point 265.0°C
Flash Point 127°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Linear Formula [(CH3)3C]2C6H2(CH3)OH
Quantity 5 kg
Beilstein 06, III, 2073
Fieser 15,113
Merck Index 15, 1550
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in toluene, soluble in methanol, ethanol, isopropanol, acetone, benzene, methyl ethyl ketone, petroleum ether,, cellowolve and most other hydrocarbon solvents
Viscosity 3.47 mm2/s (0°C)
Formula Weight 220.35
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material 2,6-Di-tert-butyl-4-methylphenol
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Di-tert-butyl-4-methylphenol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'