missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(6-Chloro-3-pyridazinylamino)ethanol, 97%

Brand:  Alfa Aesar™ H61432.06

Code : NEW

Additional Details : CAS Number : 51947-89-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15487978

  • £384.90 / 5g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

2-(6-Chloro-3-pyridazinylamino)ethanol
51947-89-8
173.6

Specifications

2-(6-Chloro-3-pyridazinylamino)ethanol
51947-89-8
97%
5g
Solid
173.6
Videos
SDS