missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,6-Bis(dodecyloxy)benzaldehyde, 97%

Brand:  Alfa Aesar™ H64087.06

Code : NEW

Additional Details : CAS Number : 1198467-57-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15461129

  • £208.70 / 5g


Estimated Shipment date
from Supplier 05-07-2018
Add to basket

Specifications

Chemical Identifiers

2,6-Bis(dodecyloxy)benzaldehyde
474.77
20848961
1198467-57-0
MFCD25372091

Specifications

2,6-Bis(dodecyloxy)benzaldehyde
Solid
474.77
20848961
5g
1198467-57-0
97%
MFCD25372091
Pale yellow
Videos
SDS