missing translation for 'onlineSavingsMsg'
Learn More

2-[(5-Nitro-2-pyridyl)oxy]ethan-1-ol, 97%, Thermo Scientific™

Catalog Number p-2697500
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
143071-39-0
C7H8N2O4
184.151
MFCD00052642
KESQFSZFUCZCEI-UHFFFAOYSA-N
2-5-nitropyridin-2-yloxy ethanol, 2-5-nitro-2-pyridyloxy ethanol, 2-5-nitropyridin-2-yl oxy ethan-1-ol, 2-5-nitro-2-pyridyl oxy ethan-1-ol, 2-2-hydroxyethoxy-5-nitropyridine, 2-5-nitropyridin-2-yl oxy ethanol, 2-5-nitro-2-pyridyloxy ethan-1-ol, acmc-209xhf, maybridge1_002682, 2-5-nitropyridin-2-yl oxyethanol
587937
2-(5-nitropyridin-2-yl)oxyethanol
C1=CC(=NC=C1[N+](=O)[O-])OCCO
This item is not returnable. View return policy
143071-39-0
C7H8N2O4
184.151
MFCD00052642
KESQFSZFUCZCEI-UHFFFAOYSA-N
2-5-nitropyridin-2-yloxy ethanol, 2-5-nitro-2-pyridyloxy ethanol, 2-5-nitropyridin-2-yl oxy ethan-1-ol, 2-5-nitro-2-pyridyl oxy ethan-1-ol, 2-2-hydroxyethoxy-5-nitropyridine, 2-5-nitropyridin-2-yl oxy ethanol, 2-5-nitro-2-pyridyloxy ethan-1-ol, acmc-209xhf, maybridge1_002682, 2-5-nitropyridin-2-yl oxyethanol
587937
2-(5-nitropyridin-2-yl)oxyethanol
C1=CC(=NC=C1[N+](=O)[O-])OCCO
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 184.15
Percent Purity 97%
Chemical Name or Material 2-[(5-Nitro-2-pyridyl)oxy]ethan-1-ol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'