missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(5-Methyl-2-thienyl)thiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62089.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15487006

  • £62.96 / 1g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

2-(5-Methyl-2-thienyl)thiazolidine-4-carboxylic acid
229.32
MFCD08559334

Specifications

2-(5-Methyl-2-thienyl)thiazolidine-4-carboxylic acid
Solid
229.32
1g
97%
MFCD08559334
Videos
SDS