missing translation for 'onlineSavingsMsg'
Learn More

2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid, 97%, Thermo Scientific™

Catalog Number 10641473
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
107367-98-6
C12H11NO3
217.224
MFCD00100005
XEWJNPORMBGGKZ-UHFFFAOYSA-N
2-5-methyl-2-phenyl-1,3-oxazol-4-yl acetic acid, 5-methyl-2-phenyl-1,3-oxazol-4-yl acetic acid, 2-5-methyl-2-phenyloxazol-4-yl acetic acid, 5-methyl-2-phenyloxazol-4-yl acetic acid, maybridge1_002118, acmc-2098vv, 2-phenyl-5-methyl-4-oxazolyiacetic acid, 2-phenyl-5-methyl-4-oxazolylacetic acid, 2-phenyl-5-methyloxazol-4-ylacetic acid, 5-methyl-2-phenyl-1,3-oxazolyl acetic acid
2775139
2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid
CC1=C(N=C(O1)C2=CC=CC=C2)CC(=O)O
This item is not returnable. View return policy
107367-98-6
C12H11NO3
217.224
MFCD00100005
XEWJNPORMBGGKZ-UHFFFAOYSA-N
2-5-methyl-2-phenyl-1,3-oxazol-4-yl acetic acid, 5-methyl-2-phenyl-1,3-oxazol-4-yl acetic acid, 2-5-methyl-2-phenyloxazol-4-yl acetic acid, 5-methyl-2-phenyloxazol-4-yl acetic acid, maybridge1_002118, acmc-2098vv, 2-phenyl-5-methyl-4-oxazolyiacetic acid, 2-phenyl-5-methyl-4-oxazolylacetic acid, 2-phenyl-5-methyloxazol-4-ylacetic acid, 5-methyl-2-phenyl-1,3-oxazolyl acetic acid
2775139
2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid
CC1=C(N=C(O1)C2=CC=CC=C2)CC(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 217.22
Percent Purity 97%
Chemical Name or Material 2-(5-Methyl-2-phenyl-1, 3-oxazol-4-yl)acetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'