missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 217.22 |
Percent Purity | 97% |
Chemical Name or Material | 2-(5-Methyl-2-phenyl-1, 3-oxazol-4-yl)acetic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'