missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(5-Methyl-2-furyl)thiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62524.03

Code : NEW

Additional Details : CAS Number : 72678-99-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15427526

  • £130.38 / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

2-(5-Methyl-2-furyl)thiazolidine-4-carboxylic acid
72678-99-0
213.25
MFCD01921870

Specifications

2-(5-Methyl-2-furyl)thiazolidine-4-carboxylic acid
72678-99-0
97%
MFCD01921870
1g
Solid
213.25
Videos
SDS