missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(5-Methyl-2-furyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64526.03

Code : NEW

Additional Details : CAS Number : 78706-11-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15471779

  • £91.90 / 1g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

2-(5-Methyl-2-furyl)benzimidazole
78706-11-3
198.23
MFCD01412910

Specifications

2-(5-Methyl-2-furyl)benzimidazole
78706-11-3
95%
MFCD01412910
1g
Solid
198.23
Videos
SDS