missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(5-Methyl-2-furyl)-5-nitrobenzimidazole, 95%

Brand:  Alfa Aesar™ H64090.06

Code : NEW

Additional Details : CAS Number : 37128-68-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15491129

  • £597.84 / 5g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

2-(5-Methyl-2-furyl)-5-nitrobenzimidazole
37128-68-0
243.22
MFCD03001005

Specifications

2-(5-Methyl-2-furyl)-5-nitrobenzimidazole
37128-68-0
95%
MFCD03001005
5g
Solid
243.22
Videos
SDS