missing translation for 'onlineSavingsMsg'
Learn More

5-Methyltryptamine, hydrochloride, 98%, Thermo Scientific™

Catalog Number p-2696319
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
C11H15N2
175.25
MFCD00012683
PYOUAIQXJALPKW-UHFFFAOYSA-O
5-methyltryptamine hydrochloride, 2-5-methyl-1h-indol-3-yl ethanamine hydrochloride, 3-2-aminoethyl-5-methylindole hydrochloride, 5-methyl-1h-indole-3-ethylamine monohydrochloride, 5-methyltryptamine hcl, 1h-indole-3-ethanamine, 5-methyl-, monohydrochloride, 2-5-methylindol-3-yl ethylamine, chloride, 5-methyltryptaminehydrochloride, 5-methyl-1h-indole-3-ethylamine hcl, pubchem24287
6917145
CC1=CC=C2NC=C(CC[NH3+])C2=C1
This item is not returnable. View return policy
C11H15N2
175.25
MFCD00012683
PYOUAIQXJALPKW-UHFFFAOYSA-O
5-methyltryptamine hydrochloride, 2-5-methyl-1h-indol-3-yl ethanamine hydrochloride, 3-2-aminoethyl-5-methylindole hydrochloride, 5-methyl-1h-indole-3-ethylamine monohydrochloride, 5-methyltryptamine hcl, 1h-indole-3-ethanamine, 5-methyl-, monohydrochloride, 2-5-methylindol-3-yl ethylamine, chloride, 5-methyltryptaminehydrochloride, 5-methyl-1h-indole-3-ethylamine hcl, pubchem24287
6917145
CC1=CC=C2NC=C(CC[NH3+])C2=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 1010-95-3
CAS Min % 97.5
CAS Max % 100.0
Melting Point 281°C to 283°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Quantity 1 g
IUPAC Name 2-(5-methyl-1H-indol-3-yl)ethanamine;hydrochloride
Formula Weight 210.71
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyltryptamine,hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'