Learn More
2,5-Dimethylthiophene 98.5%, ACROS Organics™
Brand: Acros Organics 116880250
Code : 29
Additional Details : CAS Number : 638-02-8 Weight : 0.07460kg Transport : UN number : 1993 Chem class : 1 Pack group : III
Quantity | 25mL |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
2, 5-Dimethylthiophene | |
638-02-8 | |
100.0 | |
98.5% | |
112.19 |
98.5% | |
98.0 | |
≥98.0% | |
C6H8S | |
MFCD00005452 |
Specifications
2, 5-Dimethylthiophene | |
638-02-8 | |
Authentic | |
Glass bottle | |
0.9840g/mL | |
Clear colorless to slightly yellow | |
≥98.0% | |
C6H8S | |
MFCD00005452 |
98.5% | |
98.0 | |
100.0 | |
1.5100 to 1.5140 (20°C, 589nm) | |
25mL | |
Liquid | |
98.5% | |
112.19 | |
17, 41 |