missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylpyrazine, 99%, Thermo Scientific Chemicals

Catalog Number p-119530
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
123-32-0
C6H8N2
108.14
MFCD00006147
LCZUOKDVTBMCMX-UHFFFAOYSA-N
2,5-dimethyl pyrazine, pyrazine, 2,5-dimethyl, 2,5-dimethylpiazine, 2,5-dimethyl-1,4-diazine, 2,5-dimethylparadiazine, 2,5-dimethyl-pyrazine, unii-v99y0muy1q, fema no. 3272, ccris 2929, natural
31252
2,5-dimethylpyrazine
CC1=CN=C(C=N1)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
123-32-0
C6H8N2
108.14
MFCD00006147
LCZUOKDVTBMCMX-UHFFFAOYSA-N
2,5-dimethyl pyrazine, pyrazine, 2,5-dimethyl, 2,5-dimethylpiazine, 2,5-dimethyl-1,4-diazine, 2,5-dimethylparadiazine, 2,5-dimethyl-pyrazine, unii-v99y0muy1q, fema no. 3272, ccris 2929, natural
31252
2,5-dimethylpyrazine
CC1=CN=C(C=N1)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 155°C
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4987 to 1.5007
Quantity 5 g
Beilstein 23, 96
Specific Gravity 0.99
Solubility Information Solubility in water: soluble. Other solubilities: soluble in organic solvents,miscible with ethanol,ethylether,acetone
Formula Weight 108.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 5-Dimethylpyrazine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Dimethylpyrazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'