missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylpyrazine, 99%, Thermo Scientific Chemicals

Catalog Number p-7082083
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
123-32-0
C6H8N2
108.144
MFCD00006147
LCZUOKDVTBMCMX-UHFFFAOYSA-N
2,5-dimethyl pyrazine, pyrazine, 2,5-dimethyl, 2,5-dimethylpiazine, 2,5-dimethyl-1,4-diazine, 2,5-dimethylparadiazine, 2,5-dimethyl-pyrazine, unii-v99y0muy1q, fema no. 3272, ccris 2929, natural
31252
2,5-dimethylpyrazine
CC1=CN=C(C=N1)C
This item is not returnable. View return policy
123-32-0
C6H8N2
108.144
MFCD00006147
LCZUOKDVTBMCMX-UHFFFAOYSA-N
2,5-dimethyl pyrazine, pyrazine, 2,5-dimethyl, 2,5-dimethylpiazine, 2,5-dimethyl-1,4-diazine, 2,5-dimethylparadiazine, 2,5-dimethyl-pyrazine, unii-v99y0muy1q, fema no. 3272, ccris 2929, natural
31252
2,5-dimethylpyrazine
CC1=CN=C(C=N1)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 15°C
Density 0.99
Boiling Point 154°C to 155°C
Flash Point 63°C (145°F)
Odor Pleasant
Refractive Index 1.5 00
Quantity 5 g
Beilstein 107052
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 2,5-Dimethylpyrazine
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'