missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethylfuran-3-carbonyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Density | 1.19g/mL |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 158.58 |
Percent Purity | 97% |
Chemical Name or Material | 2, 5-Dimethylfuran-3-carbonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'