missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylbenzylamine, 98%, Thermo Scientific Chemicals

Catalog Number p-2689819
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
93-48-1
C9H13N
135.21
MFCD00039780
LUJNPFWZXIGIPS-UHFFFAOYSA-N
66735
(2,5-dimethylphenyl)methanamine
CC1=CC(=C(C=C1)C)CN
This item is not returnable. View return policy
93-48-1
C9H13N
135.21
MFCD00039780
LUJNPFWZXIGIPS-UHFFFAOYSA-N
66735
(2,5-dimethylphenyl)methanamine
CC1=CC(=C(C=C1)C)CN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5350 to 1.538
Quantity 1 g
Synonym 2,5-dimethylbenzylamine, 2,5-dimethylphenyl methanamine, 1-2,5-dimethylphenyl methanamine, benzenemethanamine, 2,5-dimethyl, 2,5-dimethyl-benzylamine, 2,5-dimethylbenzenemethanamine, 2,5-dimethylphenyl methylamine, 2,5-dimethylphenyl methylamine, 2-methylamino-p-xylene, 2, 5-dimethylbenzylamine, 2,5-dimethylbenzylam
Formula Weight 135.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 5-Dimethylbenzylamine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Dimethylbenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'