missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethylbenzyl alcohol, 97+%, Thermo Scientific Chemicals
24.73 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
2,5-Dimethylbenzyl alcohol, is used as a self-indicating standard for the determination of organ lithium reagents
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,5-Dimethylbenzyl alcohol, is used as a self-indicating standard for the determination of organ lithium reagents
Solubility
Difficult in mix.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2,5-Dimethylbenzyl alcohol, is used as a self-indicating standard for the determination of organ lithium reagents
Solubility
Difficult in mix.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 40°C to 42°C |
Density | 1.011 |
Boiling Point | 232°C to 234°C |
Flash Point | 107°C (224°F) |
Refractive Index | 1.536 |
Quantity | 1 g |
Beilstein | 2243420 |
Synonym | 2,5-dimethylbenzyl alcohol, 2,5-dimethylphenyl methanol, benzenemethanol, 2,5-dimethyl, acmc-1ayay, 2,5-dimethylbenzylalcohol, benzenemethanol,2,5-dimethyl, 2,5-dimethylphenyl methanol #, 2'-oxy-1.2.4-trimethylbenzol, 2,5-dimethyl-hydroxymethylbenzene |
Solubility Information | Difficult in mix. |
Formula Weight | 136.19 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'