Learn More
2,5-Dimethylbenzoxazole 99%, ACROS Organics™
Brand: Acros Organics 116060100
Code : 29
Additional Details : CAS Number : 5676-58-4 Weight : 0.06000kg
Quantity | 10g |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
2, 5-Dimethylbenzoxazole | |
5676-58-4 | |
100.0 | |
99% | |
147.18 |
99% | |
98.5 | |
≥98.5% | |
C9H9NO | |
MFCD00005773 |
Specifications
2, 5-Dimethylbenzoxazole | |
Authentic | |
10g | |
5676-58-4 | |
100.0 | |
1.5425 to 1.5445 (20°C, 589nm) | |
≥98.5% | |
C9H9NO | |
MFCD00005773 |
1.0900g/mL | |
Clear yellow to brown | |
99% | |
98.5 | |
Glass bottle | |
Liquid | |
99% | |
147.18 | |
27, 49 |