missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylaniline, 99%, Thermo Scientific Chemicals

Catalog Number p-115392
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
95-78-3
C8H11N
121.18
MFCD00007743
VOWZNBNDMFLQGM-UHFFFAOYSA-N
2,5-xylidine, p-xylidine, 2,5-dimethylphenylamine, 2-amino-1,4-xylene, 2,5-dimethylbenzenamine, benzenamine, 2,5-dimethyl, 5-methyl-o-toluidine, 6-methyl-m-toluidine, 1-amino-2,5-dimethylbenzene, p-dimethylaniline
7259
CHEBI:518305
2,5-dimethylaniline
CC1=CC=C(C)C(N)=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
95-78-3
C8H11N
121.18
MFCD00007743
VOWZNBNDMFLQGM-UHFFFAOYSA-N
2,5-xylidine, p-xylidine, 2,5-dimethylphenylamine, 2-amino-1,4-xylene, 2,5-dimethylbenzenamine, benzenamine, 2,5-dimethyl, 5-methyl-o-toluidine, 6-methyl-m-toluidine, 1-amino-2,5-dimethylbenzene, p-dimethylaniline
7259
CHEBI:518305
2,5-dimethylaniline
CC1=CC=C(C)C(N)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 11.5°C
CAS Min % 98.5
CAS Max % 100.0
Density 0.9700g/mL
Boiling Point 218°C
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5582 to 1.5602
Assay Percent Range 98.5% min. (GC)
Specific Gravity 0.97
Linear Formula (CH3)2C6H3NH2
Beilstein 12, 1135
Merck Index 15, 10282
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether,carbon tetrachloride
Formula Weight 121.18
Percent Purity 99%
Chemical Name or Material 2, 5-Dimethylaniline, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,5-Dimethylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'