missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[((5-[(dimethylamino)methyl]-2-furyl)methyl)thio]ethan-1-amine, Maybridge™
£20.09 - £97.45
Chemical Identifiers
CAS | 66356-53-4 |
---|---|
Molecular Formula | C10H18N2OS |
Molecular Weight (g/mol) | 214.327 |
InChI Key | JFGCGQJHMUYGLU-UHFFFAOYSA-N |
Synonym | 2-2-aminoethylthio methyl-5-n,n-dimethylamino methyl furan, unii-s8w8i89w43, 2-5-dimethylamino methylfuryl thio ethylamine, 2-2-aminoethylthiomethyl-5-dimethylaminomethylfuran, 5-2-aminoethyl thiomethyl-n,n-dimethyl-2-furanmethanamine, 2-furanmethanamine, 5-2-aminoethyl thio methyl-n,n-dimethyl, 2-5-dimethylamino methyl-2-furyl methylsulfanyl ethanamine, 5-2-aminoethyl thio methyl-n,n-dimethyl-2-furanmethanamine, 2-5-dimethylamino methyl-2-furyl methyl thio ethan-1-amine |
PubChem CID | 162203 |
IUPAC Name | 2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethanamine |
SMILES | CN(C)CC1=CC=C(O1)CSCCN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10018261
|
Maybridge
SB01748DA |
1g |
£20.09
1g |
Estimated Shipment 22-02-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10342255
|
Maybridge
SB01748DE |
5g |
£59.27
5g |
Estimated Shipment 22-02-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10273132
|
Maybridge
SB01748EA |
10g |
£97.45
10g |
Estimated Shipment 22-02-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
66356-53-4 | |
214.327 | |
2-2-aminoethylthio methyl-5-n,n-dimethylamino methyl furan, unii-s8w8i89w43, 2-5-dimethylamino methylfuryl thio ethylamine, 2-2-aminoethylthiomethyl-5-dimethylaminomethylfuran, 5-2-aminoethyl thiomethyl-n,n-dimethyl-2-furanmethanamine, 2-furanmethanamine, 5-2-aminoethyl thio methyl-n,n-dimethyl, 2-5-dimethylamino methyl-2-furyl methylsulfanyl ethanamine, 5-2-aminoethyl thio methyl-n,n-dimethyl-2-furanmethanamine, 2-5-dimethylamino methyl-2-furyl methyl thio ethan-1-amine | |
2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethanamine |
C10H18N2OS | |
JFGCGQJHMUYGLU-UHFFFAOYSA-N | |
162203 | |
CN(C)CC1=CC=C(O1)CSCCN |
Specifications
2-[({5-[(Dimethylamino)methyl]-2-furyl}methyl)thio]ethan-1-amine | |
66356-53-4 | |
C10H18N2OS | |
JFGCGQJHMUYGLU-UHFFFAOYSA-N | |
2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethanamine | |
162203 | |
95% |
1g | |
95% | |
2-2-aminoethylthio methyl-5-n,n-dimethylamino methyl furan, unii-s8w8i89w43, 2-5-dimethylamino methylfuryl thio ethylamine, 2-2-aminoethylthiomethyl-5-dimethylaminomethylfuran, 5-2-aminoethyl thiomethyl-n,n-dimethyl-2-furanmethanamine, 2-furanmethanamine, 5-2-aminoethyl thio methyl-n,n-dimethyl, 2-5-dimethylamino methyl-2-furyl methylsulfanyl ethanamine, 5-2-aminoethyl thio methyl-n,n-dimethyl-2-furanmethanamine, 2-5-dimethylamino methyl-2-furyl methyl thio ethan-1-amine | |
CN(C)CC1=CC=C(O1)CSCCN | |
214.327 | |
214.33 |