missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethyl-3-hexyne-2,5-diol, 98%, Thermo Scientific™
Click to view available options
Quantity:
100 g
500 g
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | >89.0°C |
Color | White to Yellow |
Boiling Point | 121.0°C to 123.0°C (7.0 mmHg) |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Plastic bottle |
Linear Formula | HOC(CH3)2C≡CC(CH3)2OH |
Quantity | 100 g |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'