missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-2,5-hexanediol, 97%, Thermo Scientific Chemicals

Catalog Number p-118077
Click to view available options
Quantity:
5 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
110-03-2
C8H18O2
146.23
MFCD00004473
ZWNMRZQYWRLGMM-UHFFFAOYSA-N
2,5-dimethyl-2,5-hexanediol, 2,5-hexanediol, 2,5-dimethyl, unii-3pnb2s8721, 1,1,4,4-tetramethyl-1,4-butanediol, 2,5-dihydroxy-2,5-dimethylhexane, 2,5-dimethyl-hexane-2,5-diol, dimethylhexanediol, dsstox_cid_6830, dsstox_rid_78222, dsstox_gsid_26830
8031
2,5-dimethylhexane-2,5-diol
CC(C)(O)CCC(C)(C)O
This item is not returnable. View return policy
110-03-2
C8H18O2
146.23
MFCD00004473
ZWNMRZQYWRLGMM-UHFFFAOYSA-N
2,5-dimethyl-2,5-hexanediol, 2,5-hexanediol, 2,5-dimethyl, unii-3pnb2s8721, 1,1,4,4-tetramethyl-1,4-butanediol, 2,5-dihydroxy-2,5-dimethylhexane, 2,5-dimethyl-hexane-2,5-diol, dimethylhexanediol, dsstox_cid_6830, dsstox_rid_78222, dsstox_gsid_26830
8031
2,5-dimethylhexane-2,5-diol
CC(C)(O)CCC(C)(C)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Absorbance GC≥96.0%
CAS Min % 98.5
CAS Max % 100.0
Melting Point 88°C to 91°C
Color White
Boiling Point 214°C to 215°C
Flash Point 126°C
Infrared Spectrum Conforms
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula HOC(CH3)2CH2CH2C(CH3)2OH
Quantity 5 g
Beilstein 01, 492
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone,ethylene glycol,insoluble in benzene,ccl4,petroleuw ether
Formula Weight 146.23
Percent Purity 97%
Physical Form Flakes, Pellets or Prills
Chemical Name or Material 2, 5-Dimethyl-2, 5-hexanediol, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'