missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-1,3-oxazole-4-carbaldehyde, 97%, Maybridge

Brand:  Maybridge CC26804DA

Code : 31

Additional Details : CAS Number : 92901-88-7 Weight : 0.05100kg

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10691475

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

2, 5-Dimethyl-1, 3-oxazole-4-carbaldehyde
97%
125.13
97%
C6H7NO2

Specifications

2, 5-Dimethyl-1, 3-oxazole-4-carbaldehyde
92901-88-7
97%
C6H7NO2
1g
Amber Glass Bottle
97%
125.13
Videos
SDS