missing translation for 'onlineSavingsMsg'
Learn More

(2,5-Dimethyl-1,3-oxazol-4-yl)methylamine, ≥97%, Maybridge

Brand:  Maybridge CC26813DA

Code : 31

Additional Details : CAS Number : 859850-62-7 Weight : 0.05100kg Transport : UN number : 2735 Chem class : 1b Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 10670964

  • £259.00 / 1g


Estimated Shipment date
from Supplier 22-02-2018
Add to basket

Specifications

Chemical Identifiers

(2, 5-Dimethyl-1, 3-oxazol-4-yl)methylamine
97%
C6H10N2O
MFCD08271883
859850-62-7
97%
126.16

Specifications

(2, 5-Dimethyl-1, 3-oxazol-4-yl)methylamine
859850-62-7
97%
126.16
1g
97%
C6H10N2O
MFCD08271883
Videos
SDS