missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethoxytetrahydrofuran, cis + trans, 98%, Thermo Scientific Chemicals
22.73 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
Description
2,5-Dimethoxytetrahydrofuran is a tetrahydrofuran derivative that finds application in proteomics research and in organic synthesis. It is also used as an intermediate for atropine sulfate, which is an anticholinergic drug. Further, it acts as a precursor of butanediol. In addition to this, it is used as a photographic hardener and as disinfectant in formulations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,5-Dimethoxytetrahydrofuran is a tetrahydrofuran derivative that finds application in proteomics research and in organic synthesis. It is also used as an intermediate for atropine sulfate, which is an anticholinergic drug. Further, it acts as a precursor of butanediol. In addition to this, it is used as a photographic hardener and as disinfectant in formulations.
Solubility
Miscible with methanol, diethyl ether and tetrahydrofuran. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong acids.
2,5-Dimethoxytetrahydrofuran is a tetrahydrofuran derivative that finds application in proteomics research and in organic synthesis. It is also used as an intermediate for atropine sulfate, which is an anticholinergic drug. Further, it acts as a precursor of butanediol. In addition to this, it is used as a photographic hardener and as disinfectant in formulations.
Solubility
Miscible with methanol, diethyl ether and tetrahydrofuran. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications
Melting Point | -45°C |
Density | 1.02 |
Boiling Point | 145°C to 147°C |
Quantity | 100 g |
Refractive Index | 1.418 |
UN Number | UN3271 |
Beilstein | 104261 |
Solubility Information | Miscible with methanol,diethyl ether and tetrahydrofuran. Immiscible with water. |
Formula Weight | 132.16 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'