missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxyphenyl isocyanate, 99%

Product Code. 11357746
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11357746

Brand: Thermo Scientific Alfa Aesar L11381.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 56309-62-7 | C9H9NO3 | 179.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56309-62-7
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD00002006
InChI Key XNQBFHMCUNRKPM-UHFFFAOYSA-N
Synonym 2,5-dimethoxyphenyl isocyanate, 2,5-dimethoxyphenylisocyanate, acmc-1az6v, 2,5-dimethoxybenzenisocyanate, 2-isocyanato-1,4-dimethoxy-benzene, benzene, 2-isocyanato-1,4-dimethoxy-9ci
PubChem CID 4178425
IUPAC Name 2-isocyanato-1,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)OC)N=C=O

Specifications

Melting Point 38°C to 42°C
Boiling Point 152°C to 154°C (26 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN2811
Beilstein 3267303
Sensitivity Moisture sensitive
Formula Weight 179.18
Percent Purity 99%
Chemical Name or Material 2,5-Dimethoxyphenyl isocyanate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.