missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorophenylboronic acid, 97%, Thermo Scientific™

Catalog Number p-2697155
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
193353-34-3
C6H5BF2O2
157.91
KTOJGSDLJNUAEP-UHFFFAOYSA-N
2,5-difluorophenyl boronic acid, 2,5-difluorobenzeneboronic acid, 2,5-difluorophenyl boranediol, boronic acid, 2,5-difluorophenyl, 2,5-difluorophenylboronicacid, 2,5-fluorobenzene boronic acid, pubchem1820, acmc-1c1am, ksc174i6r, 2,5-difluoro phenylboronic acid
2734335
(2,5-difluorophenyl)boronic acid
B(C1=C(C=CC(=C1)F)F)(O)O
This item is not returnable. View return policy
193353-34-3
C6H5BF2O2
157.91
KTOJGSDLJNUAEP-UHFFFAOYSA-N
2,5-difluorophenyl boronic acid, 2,5-difluorobenzeneboronic acid, 2,5-difluorophenyl boranediol, boronic acid, 2,5-difluorophenyl, 2,5-difluorophenylboronicacid, 2,5-fluorobenzene boronic acid, pubchem1820, acmc-1c1am, ksc174i6r, 2,5-difluoro phenylboronic acid
2734335
(2,5-difluorophenyl)boronic acid
B(C1=C(C=CC(=C1)F)F)(O)O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 105°C to 110°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 157.91
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 5-Difluorophenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Difluorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'