missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluoronitrobenzene, 99%, ACROS Organics™

Brand:  Acros Organics 268920250

Code : 29

Additional Details : CAS Number : 364-74-9 Weight : 0.07500kg Transport : UN number : 2810 Chem class : 1 Pack group : III


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10606861

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

2, 5-Difluoronitrobenzene
98.5
99%
C6H3F2NO2
159.09
364-74-9
100.0
99%
F2C6H3NO2
MFCD00007054

Specifications

2, 5-Difluoronitrobenzene
90°C
Clear yellow
25g
364-74-9
100.0
1.5085 to 1.5105
99%
C6H3F2NO2
159.09
05, 129
1.4620g/mL
207.0°C
− 12.0°C
Authentic
98.5
Glass bottle
Liquid
99%
F2C6H3NO2
MFCD00007054
Videos
SDS