missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzyl chloride, 97%, Thermo Scientific™

Catalog Number 11372719
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
495-07-8
C7H5ClF2
162.564
MFCD01090989
INXKTZMJFPRVAY-UHFFFAOYSA-N
2,5-difluorobenzyl chloride, 2-chloromethyl-1,4-difluorobenzene, 2,5-difluorobenzylchloride, alpha-chloro-2,5-difluorotoluene, benzene, 2-chloromethyl-1,4-difluoro, 2-chloromethyl-1,4-bis fluoranyl benzene, 2-chloromethyl-1,4-difluoro-benzene, acmc-1an3f, ksc591g4f
2736950
2-(chloromethyl)-1,4-difluorobenzene
C1=CC(=C(C=C1F)CCl)F
This item is not returnable. View return policy
495-07-8
C7H5ClF2
162.564
MFCD01090989
INXKTZMJFPRVAY-UHFFFAOYSA-N
2,5-difluorobenzyl chloride, 2-chloromethyl-1,4-difluorobenzene, 2,5-difluorobenzylchloride, alpha-chloro-2,5-difluorotoluene, benzene, 2-chloromethyl-1,4-difluoro, 2-chloromethyl-1,4-bis fluoranyl benzene, 2-chloromethyl-1,4-difluoro-benzene, acmc-1an3f, ksc591g4f
2736950
2-(chloromethyl)-1,4-difluorobenzene
C1=CC(=C(C=C1F)CCl)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 5 g
UN Number UN1760
Formula Weight 162.57
Percent Purity 97%
Chemical Name or Material 2,5-Difluorobenzyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'