missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzoyl chloride, 97+%, Thermo Scientific Chemicals

Catalog Number p-7080889
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
35730-09-7
C7H3ClF2O
176.547
MFCD00009929
RLRUKKDFNWXXRT-UHFFFAOYSA-N
2,5-difluorobenzoylchloride, benzoyl chloride, 2,5-difluoro, benzoyl chloride,2,5-difluoro, acmc-1afez, 2,5-difluoro benzoyl chloride, 2,5-difluoro-benzoyl chloride, 2,5-difluorobenzoyl chloride
588082
2,5-difluorobenzoyl chloride
C1=CC(=C(C=C1F)C(=O)Cl)F
This item is not returnable. View return policy
35730-09-7
C7H3ClF2O
176.547
MFCD00009929
RLRUKKDFNWXXRT-UHFFFAOYSA-N
2,5-difluorobenzoylchloride, benzoyl chloride, 2,5-difluoro, benzoyl chloride,2,5-difluoro, acmc-1afez, 2,5-difluoro benzoyl chloride, 2,5-difluoro-benzoyl chloride, 2,5-difluorobenzoyl chloride
588082
2,5-difluorobenzoyl chloride
C1=CC(=C(C=C1F)C(=O)Cl)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.425
Boiling Point 81°C to 83°C (14 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.515
Quantity 5 g
UN Number UN3265
Beilstein 2046666
Sensitivity Moisture sensitive
Formula Weight 176.55
Percent Purity 98%
Chemical Name or Material 2,5-Difluorobenzoyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'