missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzaldehyde, 98%, ACROS Organics™

Brand:  Acros Organics 303520010

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 2646-90-4 Weight : 0.05100kg Transport : UN number : 1993 Chem class : 1 Pack group : III

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10630395

  • £12.60 / 1g


EU Stock 8
Add to basket

Specifications Description & Specifications

Chemical Identifiers

2, 5-Difluorobenzaldehyde
97.5
≥97.5%
C7H4F2O
142.11
2646-90-4
100.0
98%
FC6H3CHO
MFCD00010327

Specifications

2, 5-Difluorobenzaldehyde
Clear colorless to yellow
2646-90-4
100.0
Glass bottle
Liquid
98%
FC6H3CHO
MFCD00010327
1.3g/mL
1g
97.5
Authentic
1.4970 to 1.5010 (20°C, 589nm)
≥97.5%
C7H4F2O
142.11
Documentation

Technical Documents