missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,5-Difluoro-N-(3-fluoro-2-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H61575.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15436616

  • £79.71 / 1g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

2,5-Difluoro-N-(3-fluoro-2-methylphenyl)benzamide
C14H10F3NO
265.23
MFCD24848985

Specifications

2,5-Difluoro-N-(3-fluoro-2-methylphenyl)benzamide
97%
265.23
1g
C14H10F3NO
MFCD24848985
Videos
SDS