missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,5-Difluoro-4-nitrotoluene, 95%

Brand:  Alfa Aesar™ H63738.03

Code : NEW

Additional Details : CAS Number : 141412-60-4 Weight : 0.00100kg Transport : UN number : 2810 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15400589

  • £12.08 / 1g


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

2,5-Difluoro-4-nitrotoluene
173.12
UN2810
141412-60-4
MFCD06200905
1,4-Difluoro-2-methyl-5-nitrobenzene

Specifications

2,5-Difluoro-4-nitrotoluene
1g
Liquid
173.12
UN2810
Colorless to pale yellow
141412-60-4
95%
MFCD06200905
1,4-Difluoro-2-methyl-5-nitrobenzene
Videos
SDS