missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluoro-4-methylbenzoic acid, 95%, Thermo Scientific™

Catalog Number 15430459
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
103877-80-1
C8H6F2O2
172.13
MFCD08063230
PWWMQUUDAAWEBK-UHFFFAOYSA-N
2,5-difluoro-4-methylbenzoicacid, 2,5-difluoro-4-methyl-benzoic acid, benzoic acid,2,5-difluoro-4-methyl, pubchem4944, acmc-1brvm, 4-methyl-2,5-difluorobenzoic acid
13657451
2,5-difluoro-4-methylbenzoic acid
CC1=C(F)C=C(C(O)=O)C(F)=C1
This item is not returnable. View return policy
103877-80-1
C8H6F2O2
172.13
MFCD08063230
PWWMQUUDAAWEBK-UHFFFAOYSA-N
2,5-difluoro-4-methylbenzoicacid, 2,5-difluoro-4-methyl-benzoic acid, benzoic acid,2,5-difluoro-4-methyl, pubchem4944, acmc-1brvm, 4-methyl-2,5-difluorobenzoic acid
13657451
2,5-difluoro-4-methylbenzoic acid
CC1=C(F)C=C(C(O)=O)C(F)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Quantity 5 g
Formula Weight 172.13
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2,5-Difluoro-4-methylbenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'