missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluoro-3-nitrotoluene, 98%, Thermo Scientific™

Catalog Number 15490999
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1093758-82-7
C7H5F2NO2
173.119
MFCD11519028
JPBHCSGGXITIHO-UHFFFAOYSA-N
2,5-difluoro-3-nitrotoluene, pubchem10407, 2,5-difluoro-nitrotoluene
55274150
2,5-difluoro-1-methyl-3-nitrobenzene
CC1=CC(=CC(=C1F)[N+](=O)[O-])F
This item is not returnable. View return policy
1093758-82-7
C7H5F2NO2
173.119
MFCD11519028
JPBHCSGGXITIHO-UHFFFAOYSA-N
2,5-difluoro-3-nitrotoluene, pubchem10407, 2,5-difluoro-nitrotoluene
55274150
2,5-difluoro-1-methyl-3-nitrobenzene
CC1=CC(=CC(=C1F)[N+](=O)[O-])F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN2811
Formula Weight 173.12
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2,5-Difluoro-3-nitrotoluene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'